4-Amino-3-nitrobenzenesulfonamide 95%

Supplier: Ambeed
Warning

A179229-100MG A179229-1G A179229-250MG A179229-5G
77287-106EA 6 USD
77287-106 77287-108 77287-110 77287-112
4-Amino-3-nitrobenzenesulfonamide 95%
4-Amino-3-nitrobenzenesulphonamide

Precaución: for Research Use Only

Formula: C₆H₇N₃O₄S
MDL Number: MFCD00116612
CAS Number: 2360-19-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR