5'-Fluoro-2'-methoxyacetophenone 98%

Supplier: Ambeed
Danger

A179178-1G A179178-5G
77273-974EA 9 USD
77273-974 77273-976
5'-Fluoro-2'-methoxyacetophenone 98%
5'-Fluoro-2'-methoxyacetophenone

Precaución: for Research Use Only

Formula: C₉H₉FO₂
MDL Number: MFCD00671763
CAS Number: 445-82-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR