7-Amino-1H-indazole 95%

Supplier: Ambeed
Warning

A179150-100G A179150-100MG A179150-10G A179150-1G A179150-250MG A179150-25G A179150-5G
77236-482EA 443.93 USD
77236-482 77236-484 77236-486 77236-488 77236-490 77236-492 77236-494
7-Amino-1H-indazole 95%
7-Amino-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₇N₃
MW: 133.15 g/mol
MDL Number: MFCD00022790
CAS Number: 21443-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR