tert-Butyl 4-(3-amino-6-((1S,3S,4S)-4-((tert-butyldimethylsilyl)oxy)-3-fluorocyclohexyl)pyrazin-2-yl)-2-fluorobenzoate 97%

Supplier: Ambeed
Warning

A179101-100MG A179101-250MG
77248-212EA 18.54 USD
77248-212 77248-214
tert-Butyl 4-(3-amino-6-((1S,3S,4S)-4-((tert-butyldimethylsilyl)oxy)-3-fluorocyclohexyl)pyrazin-2-yl)-2-fluorobenzoate 97%
tert-Butyl 4-(3-amino-6-((1S,3S,4S)-4-((tert-butyldimethylsilyl)oxy)-3-fluorocyclohexyl)pyrazin-2-yl)-2-fluorobenzoate

Precaución: for Research Use Only

Formula: C₂₇H₃₉F₂N₃O₃Si
MW: 519.70 g/mol
MDL Number: MFCD30609554
CAS Number: 1715032-87-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR