1-Isobutylpiperidin-4-one 97%

Supplier: Ambeed
Warning

A179080-10G A179080-1G A179080-25G A179080-5G
77236-474EA 17 USD
77236-474 77236-476 77236-478 77236-480
1-Isobutylpiperidin-4-one 97%
1-Isobutylpiperidin-4-one

Precaución: for Research Use Only

Formula: C₉H₁₇NO
MW: 155.23 g/mol
MDL Number: MFCD00190595
CAS Number: 72544-16-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR