C-DIM 12 98%

Supplier: Ambeed
Warning

Synonyms: 3,3'-[(4-Chlorophenyl)methylene]bis[1H-indole], 3,3'-((4-Chlorophenyl)methylene)bis(1H-indole), C-DIM12

A179040-10MG A179040-1MG A179040-25MG A179040-50MG A179040-5MG
77236-464EA 100.94 USD
77236-464 77236-466 77236-468 77236-470 77236-472
C-DIM 12 98%
C-DIM 12

Precaución: for Research Use Only

Formula: C₂₃H₁₇ClN₂
MW: 356.9 g/mol
MDL Number: MFCD05625919
CAS Number: 178946-89-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR