3,4-Dimethylanisole 99%

Supplier: Ambeed
Danger

A179008-25G A179008-5G
77273-970EA 50 USD
77273-970 77273-972
3,4-Dimethylanisole 99%
3,4-Dimethylanisole

Precaución: for Research Use Only

Formula: C₉H₁₂O
MDL Number: MFCD00008396
CAS Number: 4685-47-6
UN: 1760
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR