4-Chloro-2-fluoro-5-nitrobenzoic acid 97%

Supplier: Ambeed
Warning

A178819-10G A178819-5G A178819-25G
77287-090EA 17 USD
77287-090 77287-092 77748-168
4-Chloro-2-fluoro-5-nitrobenzoic acid 97%
4-Chloro-2-fluoro-5-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃ClFNO
MDL Number: MFCD04115725
CAS Number: 35112-05-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR