4-Chloro-2-methyl-6-(trifluoromethyl)pyrimidine 97%

Supplier: Ambeed
Warning

A178589-1G A178589-5G
77287-086EA 75.19 USD
77287-086 77287-088
4-Chloro-2-methyl-6-(trifluoromethyl)pyrimidine 97%
4-Chloro-2-methyl-6-(trifluoromethyl)pyrimidine

Precaución: for Research Use Only

Formula: C₆H₄ClF₃N₂
MW: 196.5576 g/mol
MDL Number: MFCD03265399
CAS Number: 5993-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR