2,6-Dichloro-4-methylnicotinic acid 97%

Supplier: Ambeed
Warning

A178574-100G A178574-25G
77248-208EA 142 USD
77248-208 77248-210
2,6-Dichloro-4-methylnicotinic acid 97%
2,6-Dichloro-4-methylnicotinic acid

Precaución: for Research Use Only

Formula: C₇H₅Cl₂NO₂
MDL Number: MFCD02932724
CAS Number: 62774-90-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR