3-Ethoxy-4-hydroxy-5-nitrobenzaldehyde 95%

Supplier: Ambeed
Warning

A178564-1G A178564-250MG A178564-5G
77273-958EA 10 USD
77273-958 77273-960 77273-962
3-Ethoxy-4-hydroxy-5-nitrobenzaldehyde 95%
3-Ethoxy-4-hydroxy-5-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉NO₅
MDL Number: MFCD00058678
CAS Number: 178686-24-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR