3,5-Dimethyl-1H-pyrazol-4-ylamine 98%

Supplier: Ambeed
Warning

Synonyms: 3,5-Dimethyl-1H-pyrazol-4-amine

A178558-250MG A178558-1G
77287-084EA 25.75 USD
77287-084 77748-167
3,5-Dimethyl-1H-pyrazol-4-ylamine 98%
3,5-Dimethyl-1H-pyrazol-4-ylamine

Precaución: for Research Use Only

Formula: C₅H₉N₃
MW: 111.15 g/mol
MDL Number: MFCD00187666
CAS Number: 5272-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR