PS-1145 98%

Supplier: Ambeed
Warning

Synonyms: N-(6-Chloro-9H-pyrido[3,4-b]indol-8-yl)nicotinamide, N-(6-chloro-9H-pyrido[3,4-b]indol-8-yl)-3-pyridinecarboxamide, IKK Inhibitor X

A178450-10MG A178450-1MG A178450-25MG A178450-50MG A178450-5MG
77236-442EA 83.43 USD
77236-442 77236-444 77236-446 77236-448 77236-450
PS-1145 98%
PS-1145

Precaución: for Research Use Only

Formula: C₁₇H₁₁ClN₄O
MW: 322.8 g/mol
MDL Number: MFCD18089825
CAS Number: 431898-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR