7-Hydroxy-3,4-dihydroquinolin-2(1H)-one 98%

Supplier: Ambeed
Warning

Synonyms: 7-Hydroxyquinolinone, 7-Hydroxy-3,4-dihydro-2(1H)-quinolinone

A178437-100G A178437-25G A178437-500G A178437-5G A178437-10G
77273-950EA 49.44 USD
77273-950 77273-952 77273-954 77273-956 77615-500
7-Hydroxy-3,4-dihydroquinolin-2(1H)-one 98%
7-Hydroxy-3,4-dihydroquinolin-2(1H)-one

Precaución: for Research Use Only

Formula: C₉H₉NO₂
MW: 163.17 g/mol
MDL Number: MFCD06410891
CAS Number: 22246-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR