3-Acetyl-1H-pyrrole 98%
Supplier: Ambeed
|
Warning
|
A178131-10G
A178131-1G
A178131-250MG
A178131-25G
A178131-5G
77273-940EA
210.12
USD
77273-940
77273-942
77273-944
77273-946
77273-948
3-Acetyl-1H-pyrrole 98%
3-Acetyl-1H-pyrrole
Precaución: for Research Use Only
Formula:
C₆H₇NO MW: 109.13 g/mol |
MDL Number:
MFCD00067759 CAS Number: 1072-82-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...