2-Chloro-1H-purin-6(7H)-one 95%

Supplier: Ambeed
Warning

A178001-100MG A178001-250MG
77260-144EA 28.84 USD
77260-144 77260-146
2-Chloro-1H-purin-6(7H)-one 95%
2-Chloro-1H-purin-6(7H)-one

Precaución: for Research Use Only

Formula: C₅H₃ClN₄O
MW: 170.56 g/mol
MDL Number: MFCD00127838
CAS Number: 13368-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR