(±)-α-Bromophenylacetic acid 98%

Supplier: Ambeed
Danger

Synonyms: α-Bromophenylacetic Acid, DL-α-Bromophenylacetic acid

A177774-1G A177774-5G
77236-430EA 7.21 USD
77236-430 77236-432
(±)-α-Bromophenylacetic acid 98%
(±)-α-Bromophenylacetic acid

Precaución: for Research Use Only

Formula: C₈H₇BrO₂
MW: 215.0440 g/mol
MDL Number: MFCD00004206
CAS Number: 4870-65-9
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR