2',4',6'-Trihydroxyacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: Phloroacetophenone, THAP, 2-Acetylphloroglucinol

A177587-100G A177587-10G A177587-25G A177587-500G A177587-5G
77248-170EA 117.42 USD
77248-170 77248-172 77248-174 77248-176 77248-178
2',4',6'-Trihydroxyacetophenone 98%
2',4',6'-Trihydroxyacetophenone

Precaución: for Research Use Only

Formula: C₈H₈O₄
MW: 168.15 g/mol
MDL Number: MFCD00149091
CAS Number: 480-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR