6-Methoxy-1-methyl-3,4-dihydroquinolin-2(1H)-one 97%

Supplier: Ambeed
Warning

A177495-1G A177495-250MG A177495-5G
77260-128EA 81.37 USD
77260-128 77260-130 77260-132
6-Methoxy-1-methyl-3,4-dihydroquinolin-2(1H)-one 97%
6-Methoxy-1-methyl-3,4-dihydroquinolin-2(1H)-one

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO₂
MW: 191.22 g/mol
MDL Number: MFCD06656216
CAS Number: 187679-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR