2-Bromoethyl phenyl sulfide 98%

Supplier: Ambeed
Warning

A177482-100G A177482-1G A177482-25G A177482-5G
77260-120EA 382 USD
77260-120 77260-122 77260-124 77260-126
2-Bromoethyl phenyl sulfide 98%
2-Bromoethyl phenyl sulphide

Precaución: for Research Use Only

Formula: C₈H₉BrS
MDL Number: MFCD00040826
CAS Number: 4837-01-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR