4-Chloro-3-methoxybenzyl bromide 98%

Supplier: Ambeed
Danger

Synonyms: 4-(Bromomethyl)-1-chloro-2-methoxybenzene

A177209-100MG A177209-1G A177209-250MG A177209-5G
77287-042EA 59.74 USD
77287-042 77287-044 77287-046 77287-048
4-Chloro-3-methoxybenzyl bromide 98%
4-Chloro-3-methoxybenzyl bromide

Precaución: for Research Use Only

Formula: C₈H₈BrClO
MW: 235.506 g/mol
MDL Number: MFCD09907927
CAS Number: 103347-14-4
UN: 1759
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR