4-Bromo-2-(morpholin-4-ylsulfonyl)anisole 98%

Supplier: Ambeed
Warning

Synonyms: 4-((5-Bromo-2-methoxyphenyl)sulphonyl)morpholine

A177127-250MG A177127-5G
77287-038EA 31.93 USD
77287-038 77287-040
4-Bromo-2-(morpholin-4-ylsulfonyl)anisole 98%
4-Bromo-2-(morpholin-4-ylsulphonyl)anisole

Precaución: for Research Use Only

Formula: C₁₁H₁₄BrNO₄S
MW: 336.2000 g/mol
MDL Number: MFCD02043681
CAS Number: 325809-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR