(Boc-amino)-PEG3-C2-Carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2,2-Dimethyl-4-oxo-3,8,11,14-tetraoxa-5-azaheptadecan-17-oic acid, Boc-N-amido-PEG3-acid, 12-(Boc-amino)-4,7,10-trioxadodecanoic acid, 12-[(tert-Butoxycarbonyl)amino]-4,7,10-trioxadodecanoic acid

A177103-100MG A177103-10G A177103-1G A177103-250MG A177103-5G
77260-110EA 33.99 USD
77260-110 77260-112 77260-114 77260-116 77260-118
(Boc-amino)-PEG3-C2-Carboxylic acid 97%
(Boc-amino)-PEG3-C2-Carboxylic acid

Precaución: for Research Use Only

Formula: C₁₄H₂₇NO₇
MW: 321.37 g/mol
MDL Number: MFCD24539476
CAS Number: 1347750-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR