4-Bromodibenzothiophene 98%

Supplier: Ambeed
Warning

A177042-100G A177042-25G A177042-5G
77287-034EA 103 USD
77287-034 77287-036 77748-158
4-Bromodibenzothiophene 98%
4-Bromodibenzothiophene

Precaución: for Research Use Only

Formula: C₁₂H₇BrS
MDL Number: MFCD02683746
CAS Number: 97511-05-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR