4-Methyl-1H-indazole 98%
Supplier: Ambeed
|
Warning
|
A176815-10G
A176815-1G
A176815-5G
77287-018EA
170.98
USD
77287-018
77287-020
77287-022
4-Methyl-1H-indazole 98%
4-Methyl-1H-indazole
Precaución: for Research Use Only
Formula:
C₈H₈N₂ MW: 132.1600 g/mol |
MDL Number:
MFCD06200231 CAS Number: 3176-63-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...