2-Amino-4-chloro-6-(trifluoromethyl)pyrimidine 96%

Supplier: Ambeed
Warning

A176635-100G A176635-10G A176635-1G A176635-250MG A176635-25G A176635-5G
77248-148EA 525.3 USD
77248-148 77248-150 77248-152 77248-154 77248-156 77248-158
2-Amino-4-chloro-6-(trifluoromethyl)pyrimidine 96%
2-Amino-4-chloro-6-(trifluoromethyl)pyrimidine

Precaución: for Research Use Only

Formula: C₅H₃ClF₃N₃
MW: 197.546 g/mol
MDL Number: MFCD06656426
CAS Number: 16097-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR