3-Bromo-4-methoxybenzoic acid 98%

Supplier: Ambeed
Warning

A176385-100G A176385-10G A176385-1G A176385-25G A176385-5G
77236-412EA 135 USD
77236-412 77236-414 77236-416 77236-418 77236-420
3-Bromo-4-methoxybenzoic acid 98%
3-Bromo-4-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₇BrO₃
MDL Number: MFCD00020295
CAS Number: 99-58-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR