4-Bromo-2-methylbenzyl fluoride 97%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-1-(fluoromethyl)-2-methylbenzene

A176232-100MG A176232-1G A176232-250MG
77260-086EA 16.48 USD
77260-086 77260-088 77260-090
4-Bromo-2-methylbenzyl fluoride 97%
4-Bromo-2-methylbenzyl fluoride

Precaución: for Research Use Only

Formula: C₈H₈BrF
MW: 203.052 g/mol
MDL Number: MFCD22492378
CAS Number: 1824142-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR