4-(Pyrimidin-2-yl)benzaldehyde 95%

Supplier: Ambeed
Warning

A176060-250MG A176060-5G
77287-010EA 21.63 USD
77287-010 77567-604
4-(Pyrimidin-2-yl)benzaldehyde 95%
4-(Pyrimidin-2-yl)benzaldehyde

Precaución: For research use only.

Formula: C₁₁H₈N₂O
MDL Number: MFCD08056282
CAS Number: 77232-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR