IPI-549 98%

Supplier: Ambeed
Warning

Synonyms: (S)-2-Amino-N-(1-(8-((1-methyl-1H-pyrazol-4-yl)ethynyl)-1-oxo-2-phenyl-1,2-dihydroisoquinolin-3-yl)ethyl)pyrazolo[1,5-a]pyrimidine-3-carboxamide, Eganelisib, 2-amino-N-[1-[8-[2-(1-methylpyrazol-4-yl)ethynyl]-1-oxo-2-phenylisoquinolin-3-yl]ethyl]pyrazolo[1,5-a]pyrimidine-3-carboxamide

A175847-10MG A175847-1MG A175847-5MG A175847-50MG A175847-100MG A175847-250MG
77260-076EA 108.15 USD
77260-076 77260-078 77260-080 77615-472 77748-147 77748-148
IPI-549 98%
IPI-549

Precaución: for Research Use Only

Formula: C₃₀H₂₄N₈O₂
MW: 528.56 g/mol
MDL Number: MFCD30533720
CAS Number: 1693758-51-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR