N-(3-((2-((3-Fluoro-4-(4-methylpiperazin-1-yl)phenyl)amino)-7H-pyrrolo[2,3-d]pyrimidin-4-yl)oxy)phenyl)acrylamide maleate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Avitinib maleate
A175435-10MG
A175435-1MG
A175435-50MG
A175435-5MG
77236-390EA
12
USD
77236-390
77236-392
77236-394
77236-396
N-(3-((2-((3-Fluoro-4-(4-methylpiperazin-1-yl)phenyl)amino)-7H-pyrrolo[2,3-d]pyrimidin-4-yl)oxy)phenyl)acrylamide maleate 97%
N-(3-((2-((3-Fluoro-4-(4-methylpiperazin-1-yl)phenyl)amino)-7H-pyrrolo[2,3-d]pyrimidin-4-yl)oxy)phenyl)acrylamide maleate
Precaución: for Research Use Only
Formula:
C₃₀H₃₀FN₇O₆ MW: 603.6 g/mol |
MDL Number:
MFCD30345532 CAS Number: 1557268-88-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...