Dibenzofuran-1-boronic acid 97%

Supplier: Ambeed
Warning

Synonyms: Dibenzo[b,d]furan-1-ylboronic acid

A175346-10G A175346-1G A175346-25G A175346-5G
77236-382EA 58 USD
77236-382 77236-384 77236-386 77236-388
Dibenzofuran-1-boronic acid 97%
Dibenzofuran-1-boronic acid

Precaución: for Research Use Only

Formula: C₁₂H₉BO₃
MDL Number: MFCD01318978
CAS Number: 162607-19-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR