3,5-Dichloro-2,4-difluoroaniline 98%

Supplier: Ambeed
Warning

A175109-100G A175109-25G A175109-5G
77286-986EA 163 USD
77286-986 77286-988 77286-990
3,5-Dichloro-2,4-difluoroaniline 98%
3,5-Dichloro-2,4-difluoroaniline

Precaución: for Research Use Only

Formula: C₆H₃Cl₂F₂N
MDL Number: MFCD00044104
CAS Number: 83121-15-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR