3,4-Difluoro-2-methoxybenzoic acid 98%

Supplier: Ambeed
Warning

A174994-1G A174994-5G A174994-25G
77273-840EA 15.45 USD
77273-840 77273-842 77567-582
3,4-Difluoro-2-methoxybenzoic acid 98%
3,4-Difluoro-2-methoxybenzoic acid

Precaución: For research use only.

Formula: C₈H₆F₂O₃
MDL Number: MFCD03428593
CAS Number: 875664-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR