2-Bromo-1-(4-bromo-3-methoxyphenyl)ethanone 97%
Supplier: Ambeed
|
Danger
|
A174963-100MG
A174963-1G
A174963-250MG
77236-376EA
63.86
USD
77236-376
77236-378
77236-380
2-Bromo-1-(4-bromo-3-methoxyphenyl)ethanone 97%
2-Bromo-1-(4-bromo-3-methoxyphenyl)ethanone
Precaución: for Research Use Only
Formula:
C₉H₈Br₂O₂ MW: 307.96 g/mol |
MDL Number:
MFCD22491114 CAS Number: 1427431-26-2 UN: 2811 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...