Dimethyl Homoterephthalate 98%

Supplier: Ambeed
Warning

Synonyms: 4-(2-Methoxy-2-oxoethyl)benzoic acid methyl ester, Methyl 4-Methoxycarbonylphenylacetate, Methyl 4-(2-Methoxy-2-oxoethyl)benzoate, Homoterephthalic acid Dimethyl ester, 4-Methoxycarbonylphenylacetic acid methyl ester

A174604-10G A174604-1G A174604-25G A174604-5G
77286-960EA 28.84 USD
77286-960 77286-962 77286-964 77286-966
Dimethyl Homoterephthalate 98%
Dimethyl Homoterephthalate

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₄
MW: 208.21 g/mol
MDL Number: MFCD07779210
CAS Number: 52787-14-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR