(S)-tert-Butyl (1-(1,3-dioxoisoindolin-2-yl)-3-(3-fluorophenyl)propan-2-yl)carbamate 97%
Supplier: Ambeed
|
Warning
|
A174553-100MG
A174553-1G
A174553-250MG
77248-118EA
25.75
USD
77248-118
77248-120
77248-122
(S)-tert-Butyl (1-(1,3-dioxoisoindolin-2-yl)-3-(3-fluorophenyl)propan-2-yl)carbamate 97%
(S)-tert-Butyl (1-(1,3-dioxoisoindolin-2-yl)-3-(3-fluorophenyl)propan-2-yl)carbamate
Precaución: for Research Use Only
Formula:
C₂₂H₂₃FN₂O₄ MW: 398.42 g/mol |
MDL Number:
MFCD28359281 CAS Number: 1201923-48-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...