1-Methyl-4-(piperidin-4-yl)piperazine hydrochloride 97%

Supplier: Ambeed
Warning

A174479-10G A174479-250MG A174479-25G A174479-5G
77236-368EA 43.26 USD
77236-368 77236-370 77236-372 77236-374
1-Methyl-4-(piperidin-4-yl)piperazine hydrochloride 97%
1-Methyl-4-(piperidin-4-yl)piperazine hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₂₂ClN₃
MW: 219.76 g/mol
MDL Number: MFCD06801088
CAS Number: 436099-90-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR