2-(4-Bromophenyl)-2-oxoethyl acetate 97%

Supplier: Ambeed
Warning

A174420-10G A174420-5G
77236-364EA 237.93 USD
77236-364 77236-366
2-(4-Bromophenyl)-2-oxoethyl acetate 97%
2-(4-Bromophenyl)-2-oxoethyl acetate

Precaución: for Research Use Only

Formula: C₁₀H₉BrO₃
MW: 257.08 g/mol
MDL Number: MFCD00174274
CAS Number: 7500-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR