(9H-Fluoren-9-yl)methyl (1-hydroxypropan-2-yl)carbamate 95%

Supplier: Ambeed
Warning

A174135-1G A174135-5G
77260-018EA 143 USD
77260-018 77260-020
(9H-Fluoren-9-yl)methyl (1-hydroxypropan-2-yl)carbamate 95%
(9H-Fluoren-9-yl)methyl (1-hydroxypropan-2-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₈H₁₉NO₃
MW: 297.35 g/mol
MDL Number: MFCD01935769
CAS Number: 851678-69-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR