(R)-Phanephos 98%

Supplier: Ambeed
Warning

Synonyms: (R)-4,12-Bis(diphenylphosphino)-[2.2]paracyclophane, (R)-PHANEPHOS, (R)-(-)-4,12-Bis(diphenylphosphino)-[2.2]-paracyclophane

A174075-50MG A174075-100MG A174075-250MG
77248-116EA 77.25 USD
77248-116 77615-456 77746-148
(R)-Phanephos 98%
(R)-Phanephos

Precaución: for Research Use Only

Formula: C₄₀H₃₄P₂
MW: 576.646 g/mol
MDL Number: MFCD03094575
CAS Number: 364732-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR