2-Fluoro-4-iodobenzoic acid 98%

Supplier: Ambeed
Warning

A174001-10G A174001-1G A174001-250MG A174001-25G A174001-5G
77260-010EA 39 USD
77260-010 77260-012 77260-014 77260-016 77748-141
2-Fluoro-4-iodobenzoic acid 98%
2-Fluoro-4-iodobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄FIO₂
MDL Number: MFCD03094173
CAS Number: 124700-40-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR