4-((1H-1,2,4-Triazol-1-yl)methyl)aniline 98%

Supplier: Ambeed
Warning

A173794-100G A173794-25G A173794-5G
77273-802EA 153.47 USD
77273-802 77273-804 77273-806
4-((1H-1,2,4-Triazol-1-yl)methyl)aniline 98%
4-((1H-1,2,4-Triazol-1-yl)methyl)aniline

Precaución: for Research Use Only

Formula: C₉H₁₀N₄
MW: 174.21 g/mol
MDL Number: MFCD00973306
CAS Number: 119192-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR