5-(4-Methoxyphenyl)[1,3,4]thiadiazol-2-ylamine 95%

Supplier: Ambeed
Warning

A173741-1G A173741-250MG A173741-5G
77248-110EA 21 USD
77248-110 77248-112 77248-114
5-(4-Methoxyphenyl)[1,3,4]thiadiazol-2-ylamine 95%
5-(4-Methoxyphenyl)[1,3,4]thiadiazol-2-ylamine

Precaución: for Research Use Only

Formula: C₉H₉N₃OS
MDL Number: MFCD00813220
CAS Number: 1014-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR