2,2-Dimethyl-1,3-propanediol 98%

Supplier: Ambeed
Danger

Synonyms: Neopentyl Glycol, 2,2-Dimethylpropane-1,3-diol

A173726-100G A173726-25G A173726-500G A173726-1000G
77248-104EA 10 USD
77248-104 77248-106 77248-108 77550-128
2,2-Dimethyl-1,3-propanediol 98%
2,2-Dimethyl-1,3-propanediol

Precaución: for Research Use Only

Formula: C₅H₁₂O₂
MDL Number: MFCD00004685
CAS Number: 126-30-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR