4-Methyl-N-(2-(3-(morpholinomethyl)imidazo[2,1-b]thiazol-6-yl)phenyl)-2-(pyridin-3-yl)thiazole-5-carboxamide 98+%

Supplier: Ambeed
Warning

Synonyms: SRT 2104

A173533-10MG A173533-1MG A173533-50MG A173533-5MG
77236-334EA 50.47 USD
77236-334 77236-336 77236-338 77236-340
4-Methyl-N-(2-(3-(morpholinomethyl)imidazo[2,1-b]thiazol-6-yl)phenyl)-2-(pyridin-3-yl)thiazole-5-carboxamide 98+%
4-Methyl-N-(2-(3-(morpholinomethyl)imidazo[2,1-b]thiazol-6-yl)phenyl)-2-(pyridin-3-yl)thiazole-5-carboxamide

Precaución: for Research Use Only

Formula: C₂₆H₂₄N₆O₂S₂
MW: 516.64 g/mol
MDL Number: MFCD22572733
CAS Number: 1093403-33-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR