2,2,2-Trifluoro-1-(pyridin-4-yl)ethanamine dihydrochloride 95%

Supplier: Ambeed
Warning

A173327-100MG A173327-1G A173327-250MG
77273-788EA 58 USD
77273-788 77273-790 77273-792
2,2,2-Trifluoro-1-(pyridin-4-yl)ethanamine dihydrochloride 95%
2,2,2-Trifluoro-1-(pyridin-4-yl)ethanamine dihydrochloride

Precaución: for Research Use Only

Formula: C₇H₉Cl₂F₃N₂
MDL Number: MFCD31568143
CAS Number: 2153472-94-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR