4-Phenoxybenzylamine 97%

Supplier: Ambeed
Warning

A173194-100G A173194-10G A173194-1G A173194-25G
77286-920EA 227 USD
77286-920 77286-922 77286-924 77286-926
4-Phenoxybenzylamine 97%
4-Phenoxybenzylamine

Precaución: for Research Use Only

Formula: C₁₃H₁₃NO
MDL Number: MFCD01310836
CAS Number: 107622-80-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR