2-[Bis(tert-butoxycarbonyl)amino]-5-bromopyrimidine 98%

Supplier: Ambeed
Warning

Synonyms: 2-[Bis(tert-Butoxycarbonyl)amino]-5-bromopyrimidine

A173140-100G A173140-25G
77248-088EA 188.49 USD
77248-088 77248-090
2-[Bis(tert-butoxycarbonyl)amino]-5-bromopyrimidine 98%
2-[Bis(tert-butoxycarbonyl)amino]-5-bromopyrimidine

Precaución: for Research Use Only

Formula: C₁₄H₂₀BrN₃O₄
MW: 374.2300 g/mol
MDL Number: MFCD08275685
CAS Number: 209959-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR