4,4'-Dinonyl-2,2'-bipyridyl 98%

Supplier: Ambeed
Warning

A173121-1G A173121-25G A173121-5G
77286-914EA 13 USD
77286-914 77286-916 77286-918
4,4'-Dinonyl-2,2'-bipyridyl 98%
4,4'-Dinonyl-2,2'-bipyridyl

Precaución: for Research Use Only

Formula: C₂₈H₄₄N₂
MDL Number: MFCD00800915
CAS Number: 142646-58-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR